Learn More
Cole-Parmer Cole-Parmer Color-Coded Vinyl Adhesive Tape, Yellow, 1"W x 36 yd/RL; 1 RL/Pk
Catalog No. NC9780808 Shop All Cole-Parmer Products
Encompass_Preferred
Ideal for coding your labware or for electrical and pipe coding. Tough vinyl tape resists scuffs. Adhesive sticks to most surfaces. Tapes are on a 3" (7.6 cm) dia core. Order multiple roll dispenser separately.
Catalog No. NC9780808 Supplier Cole-Parmer Supplier No. 8620941
Ideal for coding your labware or for electrical and pipe coding. Tough vinyl tape resists scuffs. Adhesive sticks to most surfaces. Tapes are on a 3" (7.6 cm) dia core. Order multiple roll dispenser separately.