Learn More
Uline TAPE 3M 5413 PI FILM 1""X36YD
Catalog No. NC1719190
Encompass
3M 5413 Polyimide Film Tape - 1" x 36 yds; Resists temperature extremes from -100¿F to 500¿F. Flame, chemical and radiation resistant. Amber polyimide Kapton¿ Type (H) backing with silicone adhesive.
Catalog No. NC1719190 Supplier Uline Supplier No. S10340
3M 5413 Polyimide Film Tape - 1" x 36 yds; Resists temperature extremes from -100¿F to 500¿F. Flame, chemical and radiation resistant. Amber polyimide Kapton¿ Type (H) backing with silicone adhesive.