Learn More
RS Hughes Inc TAPE 3M 681 3IN X72YD 12/CS
SDP
Catalog No. NC1497619
Encompass_Preferred
3M Scotch 681 Box Sealing Tape 03485, 3 in x 72 yd, Clear 12/csResistant Properties: Chemical-resistant3M Number: 681Backing Material: UPVC FilmAdhesive Material: RubberColor: ClearWidth: 3 inLength: 72 ydTape Grade: Light DutyElongation: 35%Adhesion Strength: 30 oz/inTensile Strength: 25 lb/inThickness: 2.4 mil
Catalog No. NC1497619 Supplier RS Hughes Inc Supplier No. 02120003485
3M Scotch 681 Box Sealing Tape 03485, 3 in x 72 yd, Clear 12/csResistant Properties: Chemical-resistant3M Number: 681Backing Material: UPVC FilmAdhesive Material: RubberColor: ClearWidth: 3 inLength: 72 ydTape Grade: Light DutyElongation: 35%Adhesion Strength: 30 oz/inTensile Strength: 25 lb/inThickness: 2.4 mil