Learn More
MSC Tape, 3M PTFE 5490, 0.75 in, 1 Roll
Catalog No. 5000788942
Encompass
3M PTFE Film Tape 5490 utilizes an extruded polytetrafluoroethylene (PTFE) film backing and a pressure-sensitive silicone adhesive to create a smooth, non-stick surface. This tape facilitates material movement in various lay-flat applications, reduces friction, eliminates squeaks, and can act as a temporary chemical barrier due to its broad temperature range of -54°C to 260°C.
  • Non-stick PTFE film provides an exceptionally slick surface for easy clean-up.
  • Silicone adhesive ensures clean removal from various surfaces.
  • Wide operating temperature range for performance on heated machinery.
  • Extruded PTFE film backing lies flatter than typical skived PTFE tapes.
  • Excellent chemical resistance for challenging applications.
  • High strength and abrasion resistance reduces friction, rattle, and machine noise.
Catalog No. 50-007-88942 Supplier MSC Supplier No. 20715603
3M PTFE Film Tape 5490 utilizes an extruded polytetrafluoroethylene (PTFE) film backing and a pressure-sensitive silicone adhesive to create a smooth, non-stick surface. This tape facilitates material movement in various lay-flat applications, reduces friction, eliminates squeaks, and can act as a temporary chemical barrier due to its broad temperature range of -54°C to 260°C.
  • Non-stick PTFE film provides an exceptionally slick surface for easy clean-up.
  • Silicone adhesive ensures clean removal from various surfaces.
  • Wide operating temperature range for performance on heated machinery.
  • Extruded PTFE film backing lies flatter than typical skived PTFE tapes.
  • Excellent chemical resistance for challenging applications.
  • High strength and abrasion resistance reduces friction, rattle, and machine noise.