Learn More
Med Vet International 3M Transpore Medical Tape, Transparent, 1 inch X 1-1/2 Yard, Plastic, Nonsterile, 1527S-1, 100/Box
Catalog No. 502829169
Encompass_Preferred
The 3M Transpore surgical tape can be torn easily for fast and neat customization for any body part It's porous for improved breathability The firm adhesion to skin and tubing allows secure placement of devices This 3M Transpore medical tape is transparent for easy monitoring and can be handled while wearing gloves The hypoallergenic medical tape is latex-free and suited for individuals with sensitive skin
Catalog No. 50-282-9169 Supplier Med Vet International Supplier No. 6188BX
The 3M Transpore surgical tape can be torn easily for fast and neat customization for any body part It's porous for improved breathability The firm adhesion to skin and tubing allows secure placement of devices This 3M Transpore medical tape is transparent for easy monitoring and can be handled while wearing gloves The hypoallergenic medical tape is latex-free and suited for individuals with sensitive skin