Learn More
Med Vet International Medical Tape 3M Durapore White 1 inch X 10 Yard Silk-Like Cloth Nonsterile, 1538-1, 120/Case
Catalog No. 502829881
Encompass_Preferred
3M Durapore surgical tape is well-suited for securing tubes and devices Material conforms to the body letting you achieve a personalized fit for immobilizing fingers and toes and stabilizing splints Nonstretch silklike cloth retains its shape for reliable performance This hypoallergenic 3M Durapore medical tape is safe for sensitive skin Bi-directional tear for quick use Tape can be torn easily – even while wearing gloves Tape is 1 inch by 10 yards and can tear bidirectionally for swift applications
Catalog No. 50-282-9881 Supplier Med Vet International Supplier No. 5775CS
3M Durapore surgical tape is well-suited for securing tubes and devices Material conforms to the body letting you achieve a personalized fit for immobilizing fingers and toes and stabilizing splints Nonstretch silklike cloth retains its shape for reliable performance This hypoallergenic 3M Durapore medical tape is safe for sensitive skin Bi-directional tear for quick use Tape can be torn easily – even while wearing gloves Tape is 1 inch by 10 yards and can tear bidirectionally for swift applications