Learn More
Med Vet International 3M Durapore Silk-Like Cloth Medical Tape, 3in X 10 Yard, White, 1538-3, 40/Case
Catalog No. 502831849
Encompass_Preferred
Medical Tape 3M™ Durapore™ White 3 Inch X 10 Yard Silk-Like Cloth NonSterile 3M Durapore surgical tape is well-suited for securing tubes and devices Material conforms to the body letting you achieve a personalized fit for immobilizing fingers and toes and stabilizing splints Nonstretch silklike cloth retains its shape for reliable performance This hypoallergenic 3M Durapore medical tape is safe for sensitive skin Bi-directional tear for quick use White tape can be torn easily – even while wearing gloves because it doesn't stick to gloves 3 inches x 10 yards
Catalog No. 50-283-1849 Supplier Med Vet International Supplier No. 5777CS
Medical Tape 3M™ Durapore™ White 3 Inch X 10 Yard Silk-Like Cloth NonSterile 3M Durapore surgical tape is well-suited for securing tubes and devices Material conforms to the body letting you achieve a personalized fit for immobilizing fingers and toes and stabilizing splints Nonstretch silklike cloth retains its shape for reliable performance This hypoallergenic 3M Durapore medical tape is safe for sensitive skin Bi-directional tear for quick use White tape can be torn easily – even while wearing gloves because it doesn't stick to gloves 3 inches x 10 yards