Learn More
Med Vet International Medical Tape 3M Durapore White 1/2 inch X 10 Yard Silk-Like Cloth Nonsterile, 1538-0, 24/Box
Catalog No. 502830869
Encompass_Preferred
3M Durapore surgical tape is well-suited for securing high-strength applications The non-stretch silk-like cloth retains its shape for reliable performance This hypoallergenic 3M Durapore medical tape is safe for sensitive skin The material conforms to the body letting you achieve a personalized fit Bi-directional tear for quick use Because the tape won't stick to gloves, it can be torn while you are wearing gloves White tape is 1/2 inch x 10 yards
Catalog No. 50-283-0869 Supplier Med Vet International Supplier No. 5774BX
3M Durapore surgical tape is well-suited for securing high-strength applications The non-stretch silk-like cloth retains its shape for reliable performance This hypoallergenic 3M Durapore medical tape is safe for sensitive skin The material conforms to the body letting you achieve a personalized fit Bi-directional tear for quick use Because the tape won't stick to gloves, it can be torn while you are wearing gloves White tape is 1/2 inch x 10 yards