Learn More
Med Vet International Medical Tape 3M Microfoam White 4 inch X 5-1/2 Yard Elastic / Foam Nonsterile, 1528-4, 18/Case
Catalog No. 502830223
Encompass_Preferred
3M Microfoam surgical tape is conformable enough to cover challenging areas such as joints It has a gentle adhesive that doesn't irritate It is hypoallergenic and doesn't contain latex, making it ideal for sensitive skin It has water-resistant properties to keep moisture out The acrylic adhesive sets quickly The white tape is 4 inch x 5-1/2 yard Each box has 3 rolls of non-sterile medical tape
Catalog No. 50-283-0223 Supplier Med Vet International Supplier No. 5960CS
3M Microfoam surgical tape is conformable enough to cover challenging areas such as joints It has a gentle adhesive that doesn't irritate It is hypoallergenic and doesn't contain latex, making it ideal for sensitive skin It has water-resistant properties to keep moisture out The acrylic adhesive sets quickly The white tape is 4 inch x 5-1/2 yard Each box has 3 rolls of non-sterile medical tape