Learn More
Med Vet International Medical Tape 3M Micropore White 1 inch X 1-1/2 Yard Paper Nonsterile, 1530S-1, 100/Box
Catalog No. 502829167
Encompass_Preferred
The 3M Micropore medical tape is breathable to help maintain proper skin health It has a residue-free adhesive that's gentle and eases removal The adhesive is water-resistant, enabling it to adhere to damp skin It conforms easily around uneven skin and curves for convenience The latex-free paper tape is hypoallergenic, ensuring that it's gentle on most skin types This tape is known in hospitals as 3M Micropore surgical tape White paper medical tape is 1 inch x 1-1/2 yards
Catalog No. 50-282-9167 Supplier Med Vet International Supplier No. 6150BX
The 3M Micropore medical tape is breathable to help maintain proper skin health It has a residue-free adhesive that's gentle and eases removal The adhesive is water-resistant, enabling it to adhere to damp skin It conforms easily around uneven skin and curves for convenience The latex-free paper tape is hypoallergenic, ensuring that it's gentle on most skin types This tape is known in hospitals as 3M Micropore surgical tape White paper medical tape is 1 inch x 1-1/2 yards