Learn More
Med Vet International Medical Tape 3M Micropore White 2 inch X 1-1/2 Yard Paper Nonsterile, 1530S-2, 50/Box
Catalog No. 502830317
Encompass_Preferred
Hypoallergenic paper tape is gentle to the skin yet adheres well and leaves minimal adhesive residue upon removal An economical, general purpose, breathable medical tape Reliably secures dressings and devices to skin This tape is known in hospitals as 3M Micropore surgical tape White paper medical tape is 2 inches x 1-1/2 yards
Catalog No. 50-283-0317 Supplier Med Vet International Supplier No. 6018BX
Hypoallergenic paper tape is gentle to the skin yet adheres well and leaves minimal adhesive residue upon removal An economical, general purpose, breathable medical tape Reliably secures dressings and devices to skin This tape is known in hospitals as 3M Micropore surgical tape White paper medical tape is 2 inches x 1-1/2 yards