Learn More
Med Vet International McKesson Paper Medical Tape, 1/2 Inch x 10 Yard, Tan, 16-47305T, 24/Box
Catalog No. 503043788
Encompass_Preferred
Medical Tape McKesson Air Permeable Paper 1/2 Inch X 10 Yard Tan NonSterile Paper construction helps prevent irritations Non-sterile medical tape is highly breathable and holds well on damp skin Removes from the skin gently
Catalog No. 50-304-3788 Supplier Med Vet International Supplier No. 1055585BX
Medical Tape McKesson Air Permeable Paper 1/2 Inch X 10 Yard Tan NonSterile Paper construction helps prevent irritations Non-sterile medical tape is highly breathable and holds well on damp skin Removes from the skin gently