Learn More
Med Vet International Medical Tape 3M Transpore Transparent 2 inch X 1-1/2 Yard Plastic Nonsterile, 1527S-2, 250/Case
Catalog No. 502829027
Encompass_Preferred
Transparent, perforated plastic tape that tears easily, works reliably Easy, straight, bi-directional tear Easy to handle with gloves Good adhesion to skin and tubing 2 inches x 1-1/2 yards
Catalog No. 50-282-9027 Supplier Med Vet International Supplier No. 6126CS
Transparent, perforated plastic tape that tears easily, works reliably Easy, straight, bi-directional tear Easy to handle with gloves Good adhesion to skin and tubing 2 inches x 1-1/2 yards