Learn More
Med Vet International McKesson Plastic Medical Tape, 2 Inch x 10 Yard, Transparent, 16-47220, Each
Catalog No. 503045529
Encompass_Preferred
Medical Tape McKesson Air Permeable Plastic 2 Inch X 10 Yard Transparent NonSterile The non-sterile McKesson medical tape secures to the dressings, helping the healing process The tape is latex-free and hypoallergenic to avoid skin irritation This transparent surgical tape allows moisture and air to pass through, creating a comfortable fit It removes easily without residue for easy cleanup This plastic tape is water-resistant, staying secure despite sweat and water The tape tears in both directions, making it simple to apply The McKesson surgical tape measures 2 inches by 10 yards
Catalog No. 50-304-5529 Supplier Med Vet International Supplier No. 455538RL
Medical Tape McKesson Air Permeable Plastic 2 Inch X 10 Yard Transparent NonSterile The non-sterile McKesson medical tape secures to the dressings, helping the healing process The tape is latex-free and hypoallergenic to avoid skin irritation This transparent surgical tape allows moisture and air to pass through, creating a comfortable fit It removes easily without residue for easy cleanup This plastic tape is water-resistant, staying secure despite sweat and water The tape tears in both directions, making it simple to apply The McKesson surgical tape measures 2 inches by 10 yards