Learn More
Med Vet International Medical Tape 3M Transpore White White 1 inch X 10 Yard Plastic Nonsterile, 1534-1, 120/Case
Numéro de catalogue. 502830125
Encompass_Preferred
The 3M Transpore white surgical tape provides secure adhesion to skin, devices and tubing It offers bi-directional tearing and is easy to use while wearing gloves This surgical tape is porous for breathability and comfort It provides a secure hold for up to 72 hours This tape resists moisture for placement onto damp skin White medical tape is 1 inch x 10 yards
Numéro de catalogue. 50-283-0125 Fournisseur Med Vet International Code fournisseur 445278CS
Ajouter au panier
Ajouter au panier
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
The 3M Transpore white surgical tape provides secure adhesion to skin, devices and tubing It offers bi-directional tearing and is easy to use while wearing gloves This surgical tape is porous for breathability and comfort It provides a secure hold for up to 72 hours This tape resists moisture for placement onto damp skin White medical tape is 1 inch x 10 yards
missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.