Learn More
Med Vet International Medical Tape 3M Transpore White White 2 inch X 10 Yard Plastic Nonsterile, 1534-2, 60/Case
Catalog No. 502830128
Encompass_Preferred
Gentle to skin Easy, straight bi-directional tear Breathable to maintain skin integrity Good initial and long-term adhesion (72 hours) Easy to handle with gloves White medical tape is 2 inches x 10 yards Sticks well to itself (over taping) Water-resistant
Catalog No. 50-283-0128 Supplier Med Vet International Supplier No. 445279CS
Gentle to skin Easy, straight bi-directional tear Breathable to maintain skin integrity Good initial and long-term adhesion (72 hours) Easy to handle with gloves White medical tape is 2 inches x 10 yards Sticks well to itself (over taping) Water-resistant