Learn More
Med Vet International Dynarex Waterproof Adhesive Tape, 1/2" X 2.5 Yd, 288/Case
Catalog No. 502832367
Encompass_Preferred
Dynarex DynaSafety Waterproof Adhesive Tape is a high-quality tape designed to help keep wounds and dressings dry and protected from outside moisture. This Waterproof Adhesive Tape is encased in a plastic spool for convenient measuring, cutting, and application. The Tape is constructed with strong adhesive materials to firmly hold dressings in place for added security. The laminated cloth tears evenly, allowing for quick and easy sizing for a secure, custom fit. Dynarex Waterproof Adhesive Tape is ideal for first aid kits, sports, outdoor activities, and home use. Dynarex DynaSafety Waterproof Adhesive Tape is designed to effectively protect wounds from external moisture. The Tape features a strong adhesive for a secure hold and a spooled design for added convenience. 
Catalog No. 50-283-2367 Supplier Med Vet International Supplier No. 3651
Dynarex DynaSafety Waterproof Adhesive Tape is a high-quality tape designed to help keep wounds and dressings dry and protected from outside moisture. This Waterproof Adhesive Tape is encased in a plastic spool for convenient measuring, cutting, and application. The Tape is constructed with strong adhesive materials to firmly hold dressings in place for added security. The laminated cloth tears evenly, allowing for quick and easy sizing for a secure, custom fit. Dynarex Waterproof Adhesive Tape is ideal for first aid kits, sports, outdoor activities, and home use. Dynarex DynaSafety Waterproof Adhesive Tape is designed to effectively protect wounds from external moisture. The Tape features a strong adhesive for a secure hold and a spooled design for added convenience. 