Learn More
Med Vet International Waterproof Medical Tape 3M Blenderm Transparent 2 inch X 5 Yard Plastic Nonsterile, 1525-2, 6/Box
Catalog No. 502830933
Encompass_Preferred
The 3M Blenderm surgical tape is waterproof to keep out moisture and contaminants It's conformable to provide coverage in hard-to-tape areas This tape is useful for securing devices and dressings Its clear design provides transparency The medical tape is hypoallergenic to be gentle on sensitive skin 2 inches x 5 yards
Catalog No. 50-283-0933 Supplier Med Vet International Supplier No. 5760BX
The 3M Blenderm surgical tape is waterproof to keep out moisture and contaminants It's conformable to provide coverage in hard-to-tape areas This tape is useful for securing devices and dressings Its clear design provides transparency The medical tape is hypoallergenic to be gentle on sensitive skin 2 inches x 5 yards