Learn More
Med Vet International Curity™ Plastic Medical Tape, 1 Inch x 10 Yard, Transparent, 8534C, 12/Box
Catalog No. 503043760
Encompass_Preferred
Curity plastic medical tape can be used to keep IV tubing and catheters securely in place; keep devices securely positioned Easy to tear, even with gloves Lack of opacity allows you to see the underlying skin Ideal for securing IV tubing and dressings that require monitoring 2.5 centimeters x 9.1 meters
Catalog No. 50-304-3760 Supplier Med Vet International Supplier No. 696192BX
Curity plastic medical tape can be used to keep IV tubing and catheters securely in place; keep devices securely positioned Easy to tear, even with gloves Lack of opacity allows you to see the underlying skin Ideal for securing IV tubing and dressings that require monitoring 2.5 centimeters x 9.1 meters