Learn More
Med Vet International Medical Tape 3M Microfoam White 1 inch X 5-1/2 Yard Elastic / Foam Nonsterile, 1528-1, 1 Roll
Catalog No. 502830245
Encompass_Preferred
3M Microfoam surgical tape is conformable enough to cover challenging areas such as joints It has a gentle adhesive that doesn't irritate It is hypoallergenic and doesn't contain latex, making it ideal for sensitive skin It has water-resistant properties to keep moisture out The acrylic adhesive sets quickly White medical tape is 1 inch x 5-1/2 yards
Catalog No. 50-283-0245 Supplier Med Vet International Supplier No. 5959RL
3M Microfoam surgical tape is conformable enough to cover challenging areas such as joints It has a gentle adhesive that doesn't irritate It is hypoallergenic and doesn't contain latex, making it ideal for sensitive skin It has water-resistant properties to keep moisture out The acrylic adhesive sets quickly White medical tape is 1 inch x 5-1/2 yards