Learn More
Med Vet International Medical Tape 3M Microfoam White 2 inch X 5-1/2 Yard Elastic / Foam Nonsterile, 1528-2, 1 Roll
Catalog No. 502830220
Encompass_Preferred
3M Microfoam white surgical tape is conformable enough to cover challenging areas such as joints It has a gentle adhesive that doesn't irritate It has water-resistant properties to keep moisture out The acrylic adhesive sets quickly Each box has 6 rolls of 2-inch x 5-1/2 yard medical tape
Catalog No. 50-283-0220 Supplier Med Vet International Supplier No. 5961RL
3M Microfoam white surgical tape is conformable enough to cover challenging areas such as joints It has a gentle adhesive that doesn't irritate It has water-resistant properties to keep moisture out The acrylic adhesive sets quickly Each box has 6 rolls of 2-inch x 5-1/2 yard medical tape