Learn More
Med Vet International Medical Tape 3M Microfoam White 3 inch X 5-1/2 Yard Elastic / Foam Nonsterile, 1528-3, 1 Roll
Catalog No. 502830853
Encompass_Preferred
3M Microfoam surgical tape is conformable enough to cover challenging areas such as joints Gentle adhesive that doesn't irritate Water-resistant properties keep moisture out The acrylic adhesive sets quickly The white tape is 3 inches x 5-1/2 yards
Catalog No. 50-283-0853 Supplier Med Vet International Supplier No. 5947RL
3M Microfoam surgical tape is conformable enough to cover challenging areas such as joints Gentle adhesive that doesn't irritate Water-resistant properties keep moisture out The acrylic adhesive sets quickly The white tape is 3 inches x 5-1/2 yards