Learn More
Med Vet International Medical Tape 3M Micropore Tan 2 inch X 10 Yard Paper Nonsterile, 1533-2, 6/Box
Catalog No. 502830314
Encompass_Preferred
Hypoallergenic paper tape that is gentle to the skin yet adheres well and leaves minimal adhesive residue upon removal General purpose, breathable surgical tape Soft and flexible for improved comfort Tears easily to size, for quick and easy application This tape is known in hospitals as 3M Micropore surgical tape Radiolucent Tan tape is 2 inches x 10 yards
Catalog No. 50-283-0314 Supplier Med Vet International Supplier No. 5997BX
Hypoallergenic paper tape that is gentle to the skin yet adheres well and leaves minimal adhesive residue upon removal General purpose, breathable surgical tape Soft and flexible for improved comfort Tears easily to size, for quick and easy application This tape is known in hospitals as 3M Micropore surgical tape Radiolucent Tan tape is 2 inches x 10 yards