Learn More
Med Vet International McKesson Medical Tape, 3 Inch x 10 Yard, White, 16-47330, Each
Catalog No. 503043562
Encompass_Preferred
Medical Tape McKesson Air Permeable Paper 3 Inch X 10 Yard White NonSterile Paper construction is hypoallergenic to help prevent irritations Non-sterile medical tape is air and moisture permeable Removes from the skin gently 3 inches x 10 yards
Catalog No. 50-304-3562 Supplier Med Vet International Supplier No. 466889RL
Medical Tape McKesson Air Permeable Paper 3 Inch X 10 Yard White NonSterile Paper construction is hypoallergenic to help prevent irritations Non-sterile medical tape is air and moisture permeable Removes from the skin gently 3 inches x 10 yards