Learn More
Med Vet International Medical Tape 3M Transpore White White 3 inch X 10 Yard Plastic Nonsterile, 1534-3, 40/Case
Supplier:  Med Vet International 447641CS
Encompass_Preferred
The 3M Transpore white surgical tape provides secure adhesion to skin, devices and tubing It offers bi-directional tearing and is easy to use while wearing gloves This 10-yard long surgical tape is porous for breathability and comfort It provides a secure hold for up to 72 hours This tape resists moisture for placement onto damp skin The 3M Transpore medical tape is latex-free to minimize irritation Each box has 4 rolls of 3-inch tape
Catalog No. 50-282-8943