Learn More
Med Vet International Medical Tape 3M Transpore White White 1 inch X 10 Yard Plastic Nonsterile, 1534-1, 1 Roll
Catalog No. 502830126
Encompass_Preferred
The 3M Transpore white surgical tape provides secure adhesion to skin, devices and tubing It offers bi-directional tearing and is easy to use while wearing gloves This surgical tape is porous for breathability and comfort It provides a secure hold for up to 72 hours This tape resists moisture for placement onto damp skin White medical tape is 1 inch x 10 yards
Catalog No. 50-283-0126 Supplier Med Vet International Supplier No. 445278RL
The 3M Transpore white surgical tape provides secure adhesion to skin, devices and tubing It offers bi-directional tearing and is easy to use while wearing gloves This surgical tape is porous for breathability and comfort It provides a secure hold for up to 72 hours This tape resists moisture for placement onto damp skin White medical tape is 1 inch x 10 yards