Learn More
Electron Microscopy Sciences Autoclave Tape "Sterilized" Darkens and Appears, 3/4" width 60 YD
SDP
Catalog No. 501930833
Encompass_Preferred
Versatile imprinted tape indicates completion of normal steam sterilization processing. Tape accepts handwritten notations, may be used to seal and identify laboratory materials before and after processing. "Sterilized" darkens in color.
Width: 3/4"
This item is not returnable. View return policy
Catalog No. 50-193-0833 Supplier Electron Microscopy Sciences Supplier No. 7701805
Versatile imprinted tape indicates completion of normal steam sterilization processing. Tape accepts handwritten notations, may be used to seal and identify laboratory materials before and after processing. "Sterilized" darkens in color.
Width: 3/4"