Learn More
Cole-Parmer Autoclave Tape with Indicating Label; 1" W; 500 in/RL, 1 RL/Pk
Catalog No. NC9537781 Shop All Cole-Parmer Products
Encompass_Preferred
Crepe tape displays a dark colored diagonal after being autoclaved at 250F(121C) for 15 minutes. Adhesive backing sticks to glass, cloth or metal. Tape peels from surfaces easily and leaves no residue. Autoclavable.
Catalog No. NC9537781 Supplier Cole-Parmer Supplier No. 0827762
Crepe tape displays a dark colored diagonal after being autoclaved at 250F(121C) for 15 minutes. Adhesive backing sticks to glass, cloth or metal. Tape peels from surfaces easily and leaves no residue. Autoclavable.