Learn More
Grainger Tape Cartridge Brown 100 ft L 1/2 in W
Catalog No. 502752981
Encompass_Preferred
The Brady Continuous Label Roll is a 1.2" wide white polyester label roll that prints using a thermal transfer printer. It has a permanent acrylic adhesive and is wound with the adhesive side out. The roll is 660' long with a 1" core and contains 3360 labels that are 2" high. It can withstand temperatures from -40°F to 302°F. The labels are suitable for wire and cable marking, circuit board and electronic component labeling, barcode labeling, and more.
Catalog No. 50-275-2981 Supplier Grainger Supplier No. 14A534
The Brady Continuous Label Roll is a 1.2" wide white polyester label roll that prints using a thermal transfer printer. It has a permanent acrylic adhesive and is wound with the adhesive side out. The roll is 660' long with a 1" core and contains 3360 labels that are 2" high. It can withstand temperatures from -40°F to 302°F. The labels are suitable for wire and cable marking, circuit board and electronic component labeling, barcode labeling, and more.