Learn More
Grainger Tape Cartridge Brown 100 ft L 4 in W
Catalog No. 502752679
Encompass_Preferred
The Grainger 14A537 is a 14-inch diameter, 537 CFM, direct drive, aluminum exhaust fan. It features a 1/3 HP, 115V motor with thermal overload protection and sealed bearings. The fan has a galvanized steel housing and is designed for general ventilation applications. It can mount vertically or horizontally with the included wall sleeve and cap. This exhaust fan meets HVI and UL standards and comes with a 1-year warranty from Grainger.
Catalog No. 50-275-2679 Supplier Grainger Supplier No. 14A537
The Grainger 14A537 is a 14-inch diameter, 537 CFM, direct drive, aluminum exhaust fan. It features a 1/3 HP, 115V motor with thermal overload protection and sealed bearings. The fan has a galvanized steel housing and is designed for general ventilation applications. It can mount vertically or horizontally with the included wall sleeve and cap. This exhaust fan meets HVI and UL standards and comes with a 1-year warranty from Grainger.