Learn More
Grainger Tape Cartridge Clear 30 ft L
Catalog No. 502752212
Encompass_Preferred
The Brady Continuous Label Roll is a 1.2" wide white polyester label roll that prints using a thermal transfer printer. It has a permanent acrylic adhesive and is wound on a 1" core. The roll is 300 feet long and prints using Brady materials. It can be used for wire and cable marking, panel and equipment ID, barcode and other identification applications. The material is smear, scratch, UV, chemical, and heat resistant. It meets UL969 and CSA 22.2.
Catalog No. 50-275-2212 Supplier Grainger Supplier No. 14A425
The Brady Continuous Label Roll is a 1.2" wide white polyester label roll that prints using a thermal transfer printer. It has a permanent acrylic adhesive and is wound on a 1" core. The roll is 300 feet long and prints using Brady materials. It can be used for wire and cable marking, panel and equipment ID, barcode and other identification applications. The material is smear, scratch, UV, chemical, and heat resistant. It meets UL969 and CSA 22.2.