Learn More
Grainger Tape Cartridge Clear 30 ft L 3/4 in W
Catalog No. 502752214
Encompass_Preferred
The Grainger Approved Clear Cartridge Tape is 30 feet long and 3/4 inch wide. It features a clear acrylic adhesive and is designed for packaging, sealing, holding, mending, splicing, and other general purpose applications. This tape adheres well to paper, plastic, glass, wood, and metal. It can be used for indoor or short-term outdoor applications. The tape is dispensed from a plastic cartridge with serrated edge for easy tearing.
Catalog No. 50-275-2214 Supplier Grainger Supplier No. 14A428
The Grainger Approved Clear Cartridge Tape is 30 feet long and 3/4 inch wide. It features a clear acrylic adhesive and is designed for packaging, sealing, holding, mending, splicing, and other general purpose applications. This tape adheres well to paper, plastic, glass, wood, and metal. It can be used for indoor or short-term outdoor applications. The tape is dispensed from a plastic cartridge with serrated edge for easy tearing.