Learn More
Grainger Tape Cartridge Orange 50 ft L
Catalog No. 502753526
Encompass_Preferred
The Grainger Orange Duct Tape is a 50 foot roll of 2 inch wide orange duct tape. It features a natural rubber adhesive that bonds well to most surfaces including metal, plastic, glass, and masonry. The tape backing is made from polyethylene coated cloth that is water resistant, flexible, and tear resistant. This general purpose duct tape is ideal for sealing, repairing, holding, reinforcing, bundling, and identifying. It can withstand temperatures from 50 to 200 degrees Fahrenheit. The tape meets UL723 and ASTM D1000 standards.
Catalog No. 50-275-3526 Supplier Grainger Supplier No. 14A414
The Grainger Orange Duct Tape is a 50 foot roll of 2 inch wide orange duct tape. It features a natural rubber adhesive that bonds well to most surfaces including metal, plastic, glass, and masonry. The tape backing is made from polyethylene coated cloth that is water resistant, flexible, and tear resistant. This general purpose duct tape is ideal for sealing, repairing, holding, reinforcing, bundling, and identifying. It can withstand temperatures from 50 to 200 degrees Fahrenheit. The tape meets UL723 and ASTM D1000 standards.