Learn More
Grainger Tape Cartridge Purple 100 ft L 4 in W
Catalog No. 502752648
Encompass_Preferred
The Brady Continuous Label Roll is a 4 inch wide white polyester label roll that is ideal for wire and cable marking, panel labeling, equipment ID, and general purpose identification. This label roll features a permanent acrylic adhesive and comes in a length of 250 feet. The material is resistant to chemicals, abrasion, temperature extremes, and UV light. It can withstand temperatures from -40°F to 250°F. The small core size allows for more labels per roll. This versatile label roll can be used for indoor and outdoor applications.
Catalog No. 50-275-2648 Supplier Grainger Supplier No. 14A531
The Brady Continuous Label Roll is a 4 inch wide white polyester label roll that is ideal for wire and cable marking, panel labeling, equipment ID, and general purpose identification. This label roll features a permanent acrylic adhesive and comes in a length of 250 feet. The material is resistant to chemicals, abrasion, temperature extremes, and UV light. It can withstand temperatures from -40°F to 250°F. The small core size allows for more labels per roll. This versatile label roll can be used for indoor and outdoor applications.