Learn More
Grainger Tape Cartridge Purple 100 ft L
Catalog No. 502752982
Encompass_Preferred
The Grainger Approved Purple Duct Tape Cartridge is a 100 foot roll of 2 inch wide purple duct tape. It features a polyethylene coated cloth backing and a rubber adhesive. This general purpose duct tape is designed for bundling, sealing, splicing, patching, masking, and other tasks. It can be used for HVAC, maintenance, construction, DIY, and other applications. The tape has good adhesion, conformability, and tensile strength. It tears straight and resists abrasion, moisture, alkalis, acids, corrosion, and UV light. The operating temperature range is 0°F to 200°F.
Catalog No. 50-275-2982 Supplier Grainger Supplier No. 14A526
The Grainger Approved Purple Duct Tape Cartridge is a 100 foot roll of 2 inch wide purple duct tape. It features a polyethylene coated cloth backing and a rubber adhesive. This general purpose duct tape is designed for bundling, sealing, splicing, patching, masking, and other tasks. It can be used for HVAC, maintenance, construction, DIY, and other applications. The tape has good adhesion, conformability, and tensile strength. It tears straight and resists abrasion, moisture, alkalis, acids, corrosion, and UV light. The operating temperature range is 0°F to 200°F.