Learn More
Grainger Tape Cartridge Sky Blue 100 ft L
Catalog No. 502752986
Encompass_Preferred
The Grainger Sky Blue Tape Cartridge is a 100 foot long, 1 inch wide pressure-sensitive tape in a plastic cartridge dispenser. It features an aggressive rubber-based adhesive and a 3.2 mil dead soft crepe paper backing. This general purpose masking tape is designed for holding, sealing, splicing, and masking applications. It can be used for color coding, marking, and identification purposes. The tape has good adhesion properties and conforms well to irregular surfaces. It is hand-tearable and dispenses smoothly. This tape cartridge is made in the USA.
Catalog No. 50-275-2986 Supplier Grainger Supplier No. 14A418
The Grainger Sky Blue Tape Cartridge is a 100 foot long, 1 inch wide pressure-sensitive tape in a plastic cartridge dispenser. It features an aggressive rubber-based adhesive and a 3.2 mil dead soft crepe paper backing. This general purpose masking tape is designed for holding, sealing, splicing, and masking applications. It can be used for color coding, marking, and identification purposes. The tape has good adhesion properties and conforms well to irregular surfaces. It is hand-tearable and dispenses smoothly. This tape cartridge is made in the USA.