Learn More
Grainger Tape Cartridge Sky Blue 100 ft L
Catalog No. 502750443
Encompass_Preferred
The Grainger 14A409 is a 14 gauge, 3 conductor SJT power cord that is UL and CSA listed. It features a molded plug on one end and stripped wire leads on the other end, with 3 wires colored black, white, and green. The cord is 15 feet long, 300 volts, with a maximum temperature rating of 167°F. The jacket is sunlight and oil resistant thermoplastic elastomer. This general purpose power supply cord is commonly used on air compressors, power tools, machinery, and other equipment requiring an extra long, durable power cord.
Catalog No. 50-275-0443 Supplier Grainger Supplier No. 14A409
The Grainger 14A409 is a 14 gauge, 3 conductor SJT power cord that is UL and CSA listed. It features a molded plug on one end and stripped wire leads on the other end, with 3 wires colored black, white, and green. The cord is 15 feet long, 300 volts, with a maximum temperature rating of 167°F. The jacket is sunlight and oil resistant thermoplastic elastomer. This general purpose power supply cord is commonly used on air compressors, power tools, machinery, and other equipment requiring an extra long, durable power cord.