Learn More
Electron Microscopy Sciences LABeler™ Cassette of 24 mm Lab Tape, Clear with black print 26 FT
SDP
Catalog No. 501929282
Encompass_Preferred
Lab tape cassette is for use with the LABeler™ Lab Printer. Tape labels can be autoclaved, boiled, or frozen to temperatures as low as -50 deg C.
Color: Clear with black print
Catalog No. 50-192-9282 Supplier Electron Microscopy Sciences Supplier No. 7760013
Lab tape cassette is for use with the LABeler™ Lab Printer. Tape labels can be autoclaved, boiled, or frozen to temperatures as low as -50 deg C.
Color: Clear with black print