Learn More
Electron Microscopy Sciences LABeler™ Cassette of 12 mm Lab Tape, Blue with white print 26 FT
SDP
Catalog No. 501929279
Encompass_Preferred
Lab tape cassette is for use with the LABeler™ Lab Printer. Tape labels can be autoclaved, boiled, or frozen to temperatures as low as -50 deg C.
Color: Blue with white print
Catalog No. 50-192-9279 Supplier Electron Microscopy Sciences Supplier No. 7760010
Lab tape cassette is for use with the LABeler™ Lab Printer. Tape labels can be autoclaved, boiled, or frozen to temperatures as low as -50 deg C.
Color: Blue with white print