Learn More
Electron Microscopy Sciences LABeler™ Cassette of 12 mm Lab Tape, Green with white print 26 FT
SDP
Catalog No. 501929280
Encompass_Preferred
Lab tape cassette is for use with the LABeler™ Lab Printer. Tape labels can be autoclaved, boiled, or frozen to temperatures as low as -50 deg C.
Color: Green with white print
Catalog No. 50-192-9280 Supplier Electron Microscopy Sciences Supplier No. 7760011
Lab tape cassette is for use with the LABeler™ Lab Printer. Tape labels can be autoclaved, boiled, or frozen to temperatures as low as -50 deg C.
Color: Green with white print