Learn More
Med Vet International McKesson Black Cast Tape, 4 Inch x 4 Yard, 115-4A, 10/Box
Catalog No. 503044952
Encompass_Preferred
Cast Tape McKesson 4 Inch X 12 Foot Fiberglass Black Stretches to conform well to contours for a more comfortable better-fitting cast Porous weave allows the skin to breathe Water-activated resin lets you control working time according to application needs Lightweight for easy mobility
Catalog No. 50-304-4952 Supplier Med Vet International Supplier No. 733698BX
Cast Tape McKesson 4 Inch X 12 Foot Fiberglass Black Stretches to conform well to contours for a more comfortable better-fitting cast Porous weave allows the skin to breathe Water-activated resin lets you control working time according to application needs Lightweight for easy mobility