Learn More
Med Vet International McKesson Blue Cast Tape, 2 Inch x 4 Yard, 115-2B, 10/Box
Catalog No. 503044566
Encompass_Preferred
Cast Tape McKesson 2 Inch X 12 Foot Fiberglass Blue Stretches to conform well to contours for a more comfortable better-fitting cast Porous weave allows the skin to breathe Water-activated resin lets you control working time according to application needs Lightweight for easy mobility
Catalog No. 50-304-4566 Supplier Med Vet International Supplier No. 733685BX
Cast Tape McKesson 2 Inch X 12 Foot Fiberglass Blue Stretches to conform well to contours for a more comfortable better-fitting cast Porous weave allows the skin to breathe Water-activated resin lets you control working time according to application needs Lightweight for easy mobility