Learn More
Med Vet International McKesson Blue Cast Tape, 3 Inch x 4 Yard, 115-3B, 10/Box
Catalog No. 503043589
Encompass_Preferred
Cast Tape McKesson 3 Inch X 12 Foot Fiberglass Blue Water-activated resin lets you control working time according to application needs Lightweight for easy mobility Stretches to conform well to contours for a more comfortable better-fitting cast Porous weave allows the skin to breathe Available in various colors and sizes to maximize satisfaction
Catalog No. 50-304-3589 Supplier Med Vet International Supplier No. 733692BX
Cast Tape McKesson 3 Inch X 12 Foot Fiberglass Blue Water-activated resin lets you control working time according to application needs Lightweight for easy mobility Stretches to conform well to contours for a more comfortable better-fitting cast Porous weave allows the skin to breathe Available in various colors and sizes to maximize satisfaction