Learn More
Med Vet International McKesson Cast Tape, Green, 3 Inch x 4 Yard, 115-3G, 10/Box
Catalog No. 503044499
Encompass_Preferred
Cast Tape McKesson 3 Inch X 12 Foot Fiberglass Green Stretches to conform well to contours for a more comfortable better-fitting cast Porous weave allows the skin to breathe Water-activated resin lets you control working time according to application needs Lightweight for easy mobility
Catalog No. 50-304-4499 Supplier Med Vet International Supplier No. 733693BX
Cast Tape McKesson 3 Inch X 12 Foot Fiberglass Green Stretches to conform well to contours for a more comfortable better-fitting cast Porous weave allows the skin to breathe Water-activated resin lets you control working time according to application needs Lightweight for easy mobility