Learn More
Med Vet International Cast Tape 3M Scotchcast Plus 3 inch X 12 Foot Fiberglass / Resin Blue, 82003B, 1 Roll
Catalog No. 502829787
Encompass_Preferred
Exposure to moisture or water initiates a chemical reaction which causes the tape to become rigid Finished casts produced from Scotchcast Plus casting tape are lightweight, strong, radiolucent, and porous Casting tape contains a lubricating agent in the resin to provide a slippery feel when exposed to water Fabric impregnated with polyurethane resin 3 inches x 12 feet
Catalog No. 50-282-9787 Supplier Med Vet International Supplier No. 198266RL
Exposure to moisture or water initiates a chemical reaction which causes the tape to become rigid Finished casts produced from Scotchcast Plus casting tape are lightweight, strong, radiolucent, and porous Casting tape contains a lubricating agent in the resin to provide a slippery feel when exposed to water Fabric impregnated with polyurethane resin 3 inches x 12 feet