Learn More
Med Vet International Cast Tape 3M Scotchcast Plus 3 inch X 12 Foot Fiberglass / Resin Purple, 82003U, Each
Catalog No. 502831379
Encompass_Preferred
Water-activated resin lets you control working time according to application needs High and early strength allows for weight bearing in 20 minutes and provides a durable cast with less chance of breakdown and the need for costly repairs or replacements Lightweight for easy mobility Stretches to conform well to contours for a more comfortable, better fitting cast Porous weave allows the skin to breathe 3 inches x 12 feet
Catalog No. 50-283-1379 Supplier Med Vet International Supplier No. 199969EA
Water-activated resin lets you control working time according to application needs High and early strength allows for weight bearing in 20 minutes and provides a durable cast with less chance of breakdown and the need for costly repairs or replacements Lightweight for easy mobility Stretches to conform well to contours for a more comfortable, better fitting cast Porous weave allows the skin to breathe 3 inches x 12 feet