Learn More
Med Vet International McKesson White Cast Tape, 3 Inch x 4 Yard, 115-3, Each
Catalog No. 503045053
Encompass_Preferred
Cast Tape McKesson 3 Inch X 12 Foot Fiberglass White Stretches to conform well to contours for a more comfortable better-fitting cast Porous weave allows the skin to breathe Water-activated resin lets you control working time according to application needs Lightweight for easy mobility 3 inches x 4 yards
Catalog No. 50-304-5053 Supplier Med Vet International Supplier No. 733690EA
Cast Tape McKesson 3 Inch X 12 Foot Fiberglass White Stretches to conform well to contours for a more comfortable better-fitting cast Porous weave allows the skin to breathe Water-activated resin lets you control working time according to application needs Lightweight for easy mobility 3 inches x 4 yards