Learn More
Med Vet International McKesson White Cast Tape, 4 Inch x 4 Yard, 115-4, 10/Box
Catalog No. 503044570
Encompass_Preferred
Cast Tape McKesson 4 Inch X 12 Foot Fiberglass White Stretches to conform well to contours for a more comfortable better-fitting cast Porous weave allows the skin to breathe Water-activated resin lets you control working time according to application needs Lightweight for easy mobility
Catalog No. 50-304-4570 Supplier Med Vet International Supplier No. 733697BX
Cast Tape McKesson 4 Inch X 12 Foot Fiberglass White Stretches to conform well to contours for a more comfortable better-fitting cast Porous weave allows the skin to breathe Water-activated resin lets you control working time according to application needs Lightweight for easy mobility