Learn More
Grainger Tape Clear 50 ft L 8 in W
Catalog No. 502749881
Encompass_Preferred
The Brady Continuous Label Roll is an 8 inch wide white polyester label roll that prints using a thermal transfer printer. It has a permanent acrylic adhesive and is designed for wire and cable marking, panel labeling, equipment ID, and general purpose identification. The roll is 8 inches wide by 300 feet long. It has a service temperature range of -40°F to 302°F and is suitable for indoor and outdoor use. The polyester material and acrylic adhesive provide good chemical and abrasion resistance.
Catalog No. 50-274-9881 Supplier Grainger Supplier No. 14A590
The Brady Continuous Label Roll is an 8 inch wide white polyester label roll that prints using a thermal transfer printer. It has a permanent acrylic adhesive and is designed for wire and cable marking, panel labeling, equipment ID, and general purpose identification. The roll is 8 inches wide by 300 feet long. It has a service temperature range of -40°F to 302°F and is suitable for indoor and outdoor use. The polyester material and acrylic adhesive provide good chemical and abrasion resistance.