Learn More
Grainger Tape Clear 6 in W 50 ft L
Catalog No. 502750186
Encompass_Preferred
The Brady Continuous Label Roll is a 6 inch wide white polyester label roll that is ideal for wire and cable marking, panel labeling, equipment ID, and general purpose identification. This label roll features a permanent acrylic adhesive and comes in a length of 25 feet. The material is resistant to chemicals, abrasion, and temperatures up to 250°F. It can be used with the Brady label printers for easy custom printed labels. This versatile label roll works on a variety of surfaces including plastic, metal, rubber, glass, and more. With high performance and durability, this Brady label roll is great for creating clear and long-lasting labels in industrial environments.
Catalog No. 50-275-0186 Supplier Grainger Supplier No. 14A589
The Brady Continuous Label Roll is a 6 inch wide white polyester label roll that is ideal for wire and cable marking, panel labeling, equipment ID, and general purpose identification. This label roll features a permanent acrylic adhesive and comes in a length of 25 feet. The material is resistant to chemicals, abrasion, and temperatures up to 250°F. It can be used with the Brady label printers for easy custom printed labels. This versatile label roll works on a variety of surfaces including plastic, metal, rubber, glass, and more. With high performance and durability, this Brady label roll is great for creating clear and long-lasting labels in industrial environments.